Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Species: |
Human | |||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
|||||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 607838 | |||||||||||||||||||||||||||||||||||||||||
Literature Links: |
GNPTG PubMed Links | |||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | ||||||
---|---|---|---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | ||||||
EAS
|
African American - Not Available | YRI (Yoruba)
|
||||||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | ||||||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | ||||||
EUR
|
||||||||
AMR
|
GNPTG - N-acetylglucosamine-1-phosphate transferase gamma subunit | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_032520.4 | 4334 | Intron | NP_115909.1 | |||
XM_017023782.1 | 4334 | Intron | XP_016879271.1 | |||
XM_017023783.1 | 4334 | Intron | XP_016879272.1 |
UNKL - unkempt family like zinc finger | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001037125.3 | 4334 | Intron | NP_001032202.1 | |||
NM_001193388.3 | 4334 | UTR 3 | NP_001180317.2 | |||
NM_001193389.1 | 4334 | UTR 3 | NP_001180318.1 | |||
NM_001276414.1 | 4334 | UTR 3 | NP_001263343.1 | |||
XM_005255505.1 | 4334 | UTR 3 | XP_005255562.1 | |||
XM_011522610.1 | 4334 | UTR 3 | XP_011520912.1 | |||
XM_011522611.1 | 4334 | UTR 3 | XP_011520913.1 | |||
XM_011522612.1 | 4334 | UTR 3 | XP_011520914.1 | |||
XM_011522613.1 | 4334 | Intron | XP_011520915.1 | |||
XM_017023567.1 | 4334 | Intron | XP_016879056.1 | |||
XM_017023568.1 | 4334 | UTR 3 | XP_016879057.1 | |||
XM_017023569.1 | 4334 | UTR 3 | XP_016879058.1 |
Set Membership: |
HapMap |