Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Species: |
Human | ||||||||||||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 604418 | ||||||||||||||||||||||||||||||||||||||||||||||||||
Literature Links: |
GJB6 PubMed Links | ||||||||||||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | ||||||
---|---|---|---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU)
|
||||||
EAS
|
African American - Not Available | YRI (Yoruba)
|
||||||
SAS
|
Chinese - Not Available | JPT (Japanese)
|
||||||
AFR
|
Japanese - Not Available | CHB (Han Chinese)
|
||||||
EUR
|
||||||||
AMR
|
GJB6 - gap junction protein beta 6 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001110219.2 | 1436 | UTR 3 | NP_001103689.1 | |||
NM_001110220.2 | 1436 | UTR 3 | NP_001103690.1 | |||
NM_001110221.2 | 1436 | UTR 3 | NP_001103691.1 | |||
NM_006783.4 | 1436 | UTR 3 | NP_006774.2 | |||
XM_017020357.1 | 1436 | UTR 3 | XP_016875846.1 | |||
XM_017020358.1 | 1436 | UTR 3 | XP_016875847.1 | |||
XM_017020359.1 | 1436 | UTR 3 | XP_016875848.1 | |||
XM_017020360.1 | 1436 | UTR 3 | XP_016875849.1 |
Set Membership: |
HapMap |