Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
52 submissions
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 176806 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
PTGER3 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
PTGER3 - prostaglandin E receptor 3 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001126044.1 | Intron | NP_001119516.1 | ||||
NM_198714.1 | Intron | NP_942007.1 | ||||
NM_198715.2 | Intron | NP_942008.1 | ||||
NM_198716.1 | Intron | NP_942009.1 | ||||
NM_198717.1 | Intron | NP_942010.1 | ||||
NM_198718.1 | Intron | NP_942011.1 | ||||
NM_198719.1 | Intron | NP_942012.1 | ||||
XM_011541810.2 | Intron | XP_011540112.1 |