Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 600266 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
SLC11A1 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
SLC11A1 - solute carrier family 11 member 1 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_000578.3 | Intron | NP_000569.3 | ||||
XM_005246793.3 | Intron | XP_005246850.1 | ||||
XM_005246794.3 | Intron | XP_005246851.1 | ||||
XM_006712709.3 | Intron | XP_006712772.1 | ||||
XM_006712710.3 | Intron | XP_006712773.1 | ||||
XM_006712711.3 | Intron | XP_006712774.1 | ||||
XM_011511684.2 | Intron | XP_011509986.1 | ||||
XM_011511685.2 | Intron | XP_011509987.1 | ||||
XM_017004765.1 | Intron | XP_016860254.1 | ||||
XM_017004766.1 | Intron | XP_016860255.1 | ||||
XM_017004767.1 | Intron | XP_016860256.1 |