Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Species: |
Human | |||||||||||||||||||||||
dbSNP Submissions: |
NA
|
|||||||||||||||||||||||
Phenotype: |
MIM: 612845 | |||||||||||||||||||||||
Literature Links: |
SENP5 PubMed Links | |||||||||||||||||||||||
Allele Nomenclature: |
||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR - Not Available | |||||
AMR - Not Available |
SENP5 - SUMO1/sentrin specific peptidase 5 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001308045.1 | Intron | NP_001294974.1 | ||||
NM_152699.4 | Intron | NP_689912.2 | ||||
XM_005269310.2 | Intron | XP_005269367.1 | ||||
XM_005269312.2 | Intron | XP_005269369.1 | ||||
XM_011512544.2 | Intron | XP_011510846.1 | ||||
XM_011512545.2 | Intron | XP_011510847.1 | ||||
XM_017005870.1 | Intron | XP_016861359.1 |