Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Species: |
Human | ||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 603442 | ||||||||||||||||||||||||||||||||
Literature Links: |
CDKL2 PubMed Links | ||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU)
|
|||
EAS - Not Available | African American - Not Available | YRI (Yoruba)
|
|||
SAS - Not Available | Chinese - Not Available | JPT (Japanese)
|
|||
AFR - Not Available | Japanese - Not Available | CHB (Han Chinese)
|
|||
EUR - Not Available | |||||
AMR - Not Available |
CDKL2 - cyclin dependent kinase like 2 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
XM_005263335.2 | 2038 | UTR 3 | XP_005263392.1 | |||
XM_006714406.2 | 2038 | UTR 3 | XP_006714469.1 | |||
XM_017008809.1 | 2038 | UTR 3 | XP_016864298.1 | |||
XM_017008810.1 | 2038 | UTR 3 | XP_016864299.1 | |||
XM_017008811.1 | 2038 | UTR 3 | XP_016864300.1 |
Set Membership: |
HapMap |