Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Species: |
Human | |||||||||||||||||||||||
dbSNP Submissions: |
NA
|
|||||||||||||||||||||||
Phenotype: |
MIM: 182134 | |||||||||||||||||||||||
Literature Links: |
HTR1F PubMed Links | |||||||||||||||||||||||
Allele Nomenclature: |
||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR - Not Available | |||||
AMR - Not Available |
HTR1F - 5-hydroxytryptamine receptor 1F | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_000866.4 | Intron | NP_000857.1 | ||||
NM_001322208.1 | Intron | NP_001309137.1 | ||||
NM_001322209.1 | Intron | NP_001309138.1 | ||||
NM_001322210.1 | Intron | NP_001309139.1 | ||||
XM_005264751.3 | Intron | XP_005264808.1 | ||||
XM_011533664.2 | Intron | XP_011531966.1 |