Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Species: |
Human | ||||||||||||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 615606 | ||||||||||||||||||||||||||||||||||||||||||||||||||
Literature Links: |
BTNL8 PubMed Links | ||||||||||||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | ||||||
---|---|---|---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU)
|
||||||
EAS
|
African American - Not Available | YRI (Yoruba)
|
||||||
SAS
|
Chinese - Not Available | JPT (Japanese)
|
||||||
AFR
|
Japanese - Not Available | CHB (Han Chinese)
|
||||||
EUR
|
||||||||
AMR
|
BTNL8 - butyrophilin like 8 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001040462.2 | 297 | Intron | NP_001035552.1 | |||
NM_001159707.1 | 297 | Intron | NP_001153179.1 | |||
NM_001159708.1 | 297 | Intron | NP_001153180.1 | |||
NM_001159709.1 | 297 | UTR 5 | NP_001153181.1 | |||
NM_001159710.1 | 297 | Intron | NP_001153182.1 | |||
NM_024850.2 | 297 | Intron | NP_079126.1 | |||
XM_011534649.1 | 297 | Intron | XP_011532951.1 | |||
XM_011534650.2 | 297 | Intron | XP_011532952.1 | |||
XM_011534651.2 | 297 | Intron | XP_011532953.1 | |||
XM_011534652.2 | 297 | Intron | XP_011532954.1 | |||
XM_011534653.2 | 297 | Intron | XP_011532955.1 |
Set Membership: |
HapMap |