Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Species: |
Human | |||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
|||||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 610982 | |||||||||||||||||||||||||||||||||||||||||
Literature Links: |
INF2 PubMed Links | |||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | ||||||
---|---|---|---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | ||||||
EAS
|
African American - Not Available | YRI (Yoruba)
|
||||||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | ||||||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | ||||||
EUR
|
||||||||
AMR
|
INF2 - inverted formin, FH2 and WH2 domain containing | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001031714.3 | 21 | Intron | NP_001026884.3 | |||
NM_022489.3 | 21 | Intron | NP_071934.3 | |||
NM_032714.2 | 21 | Intron | NP_116103.1 | |||
XM_005268004.4 | 21 | UTR 5 | XP_005268061.1 | |||
XM_005268005.4 | 21 | UTR 5 | XP_005268062.1 | |||
XM_017021595.1 | 21 | UTR 5 | XP_016877084.1 |