Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Species: |
Human | ||||||||||||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 611196 | ||||||||||||||||||||||||||||||||||||||||||||||||||
Literature Links: |
ZMIZ2 PubMed Links | ||||||||||||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | ||||||
---|---|---|---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU)
|
||||||
EAS
|
African American - Not Available | YRI (Yoruba)
|
||||||
SAS
|
Chinese - Not Available | JPT (Japanese)
|
||||||
AFR
|
Japanese - Not Available | CHB (Han Chinese)
|
||||||
EUR
|
||||||||
AMR
|
ZMIZ2 - zinc finger MIZ-type containing 2 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001300959.1 | Intron | NP_001287888.1 | ||||
NM_031449.3 | Intron | NP_113637.3 | ||||
NM_174929.2 | Intron | NP_777589.2 | ||||
XM_005249866.2 | Intron | XP_005249923.1 | ||||
XM_005249867.4 | Intron | XP_005249924.1 | ||||
XM_005249868.4 | Intron | XP_005249925.1 | ||||
XM_005249869.2 | Intron | XP_005249926.1 | ||||
XM_005249870.2 | Intron | XP_005249927.1 | ||||
XM_005249871.2 | Intron | XP_005249928.1 | ||||
XM_005249872.2 | Intron | XP_005249929.1 | ||||
XM_005249873.2 | Intron | XP_005249930.1 | ||||
XM_011515566.1 | Intron | XP_011513868.1 | ||||
XM_017012674.1 | Intron | XP_016868163.1 |