Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Species: |
Human | ||||||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 604902 MIM: 610423 | ||||||||||||||||||||||||||||||||||||||||||||
Literature Links: |
BRF1 PubMed Links | ||||||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | ||||||
---|---|---|---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | ||||||
EAS
|
African American - Not Available | YRI (Yoruba)
|
||||||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | ||||||
AFR
|
Japanese - Not Available | JPT (Japanese)
|
||||||
EUR
|
||||||||
AMR
|
BRF1 - BRF1, RNA polymerase III transcription initiation factor 90 kDa subunit | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
PACS2 - phosphofurin acidic cluster sorting protein 2 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001100913.2 | Intron | NP_001094383.2 | ||||
NM_001243127.2 | Intron | NP_001230056.1 | ||||
NM_015197.3 | Intron | NP_056012.2 | ||||
XM_006720090.1 | Intron | XP_006720153.1 | ||||
XM_006720091.1 | Intron | XP_006720154.1 | ||||
XM_006720092.2 | Intron | XP_006720155.1 | ||||
XM_017021105.1 | Intron | XP_016876594.1 | ||||
XM_017021106.1 | Intron | XP_016876595.1 | ||||
XM_017021107.1 | Intron | XP_016876596.1 | ||||
XM_017021108.1 | Intron | XP_016876597.1 | ||||
XM_017021109.1 | Intron | XP_016876598.1 | ||||
XM_017021110.1 | Intron | XP_016876599.1 | ||||
XM_017021111.1 | Intron | XP_016876600.1 | ||||
XM_017021112.1 | Intron | XP_016876601.1 |