Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 186360 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
ANXA7 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
ANXA7 - annexin A7 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001156.4 | 1431 | Silent Mutation | GTA,GTG | V,V 430 | NP_001147.1 | |
NM_001320879.1 | 1431 | Silent Mutation | GTA,GTG | V,V 390 | NP_001307808.1 | |
NM_001320880.1 | 1431 | Silent Mutation | GTA,GTG | V,V 412 | NP_001307809.1 | |
NM_004034.3 | 1431 | Silent Mutation | GTA,GTG | V,V 452 | NP_004025.1 | |
XM_017016162.1 | 1431 | Silent Mutation | GTA,GTG | V,V 452 | XP_016871651.1 | |
XM_017016163.1 | 1431 | Silent Mutation | GTA,GTG | V,V 430 | XP_016871652.1 |