Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
TCAGGCCTTCTGACTGACTTCTCAA[C/T]GAAGCACGGACTGCCAATGCCAGCA
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||
Phenotype: |
|||||||||||||||||||||
Literature Links: |
TMEM26 PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
TMEM26 - transmembrane protein 26 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_178505.7 | 1311 | Silent Mutation | TCA,TCG | S,S 314 | NP_848600.2 | |
XM_011539450.2 | 1311 | Intron | XP_011537752.1 | |||
XM_011539451.1 | 1311 | Intron | XP_011537753.1 | |||
XM_017015829.1 | 1311 | Intron | XP_016871318.1 |