Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
AAAATGCTGGGACTGAGGAAATAAA[A/G]AATGAAATAAATGGAAATTGGATTT
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 608610 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
PDCD4 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
PDCD4 - programmed cell death 4 (neoplastic transformation inhibitor) | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001199492.1 | 355 | Silent Mutation | AAA,AAG | K,K 23 | NP_001186421.1 | |
NM_014456.4 | 355 | Silent Mutation | AAA,AAG | K,K 37 | NP_055271.2 | |
NM_145341.3 | 355 | Silent Mutation | AAA,AAG | K,K 26 | NP_663314.1 |