Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
AGACCAGTGCCCTGCCCACTGGCTT[C/T]GGGGAGCTCGAGGTGCTGGCTGTGG
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 616103 MIM: 600342 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
LRIT1 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
LRIT1 - leucine rich repeat, Ig-like and transmembrane domains 1 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
RGR - retinal G protein coupled receptor | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001012720.1 | 71 | Silent Mutation | TTC,TTT | F,F 11 | NP_001012738.1 | |
NM_001012722.1 | 71 | Silent Mutation | TTC,TTT | F,F 11 | NP_001012740.1 | |
NM_002921.3 | 71 | Silent Mutation | TTC,TTT | F,F 11 | NP_002912.2 |