Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
CTGACAGGTATGTCTTCTACAGTTT[G/T]AGAGAAGGGCCAGAATCCATACAGC
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 610139 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
ST8SIA6 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
ST8SIA6 - ST8 alpha-N-acetyl-neuraminide alpha-2,8-sialyltransferase 6 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001004470.1 | 1034 | Missense Mutation | AAA,CAA | K,Q 348 | NP_001004470.1 | |
XM_017016174.1 | 1034 | Missense Mutation | AAA,CAA | K,Q 318 | XP_016871663.1 | |
XM_017016175.1 | 1034 | Intron | XP_016871664.1 |