Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
TGATGATCTTTAATTGCTGCCTGTA[C/T]GGTGCAATAATACCAATATCAGAGG
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||
Phenotype: |
MIM: 601810 | ||||||||||||||||||||
Literature Links: |
DNA2 PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
DNA2 - DNA replication helicase/nuclease 2 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001080449.2 | 2324 | Silent Mutation | CCA,CCG | P,P 942 | NP_001073918.2 | |
XM_006717680.3 | 2324 | Silent Mutation | CCA,CCG | P,P 972 | XP_006717743.1 | |
XM_011539417.1 | 2324 | Silent Mutation | CCA,CCG | P,P 582 | XP_011537719.1 | |
XM_017015799.1 | 2324 | Silent Mutation | CCA,CCG | P,P 718 | XP_016871288.1 |