Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
ACAGCATCACTCACCTTGTTGTACA[C/T]GTTAAACCATTCAGGATGGTGGTCC
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 126090 MIM: 603729 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
PCBD1 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
PCBD1 - pterin-4 alpha-carbinolamine dehydratase 1 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_000281.3 | 604 | Missense Mutation | ATG,GTG | M,V 69 | NP_000272.1 | |
NM_001289797.1 | 604 | Missense Mutation | ATG,GTG | M,V 20 | NP_001276726.1 | |
NM_001323004.1 | 604 | Missense Mutation | ATG,GTG | M,V 69 | NP_001309933.1 |
SGPL1 - sphingosine-1-phosphate lyase 1 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |