Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
ATTTTTCTGGAAGGAAAAGAACTTA[C/T]GTCCATGTTAAATGGTATTCCTATT
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||
Phenotype: |
MIM: 194522 | ||||||||||||||||||||
Literature Links: |
ZNF33B PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
ZNF33B - zinc finger protein 33B | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001305033.1 | Intron | NP_001291962.1 | ||||
NM_001305035.1 | Intron | NP_001291964.1 | ||||
NM_001305036.1 | Intron | NP_001291965.1 | ||||
NM_001305037.1 | Intron | NP_001291966.1 | ||||
NM_001305038.1 | Intron | NP_001291967.1 | ||||
NM_001305039.1 | Intron | NP_001291968.1 | ||||
NM_001305040.1 | Intron | NP_001291969.1 | ||||
NM_006955.2 | Intron | NP_008886.1 |