Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
GTATGTCAAGAGCCCCACCTTCCTC[A/G]GGGGCACCTCCAGCCTCAACAGCAC
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 607185 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
SEC24C PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
SEC24C - SEC24 homolog C, COPII coat complex component | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_004922.3 | 587 | Silent Mutation | TCA,TCG | S,S 66 | NP_004913.2 | |
NM_198597.2 | 587 | Silent Mutation | TCA,TCG | S,S 66 | NP_940999.1 | |
XM_011540380.2 | 587 | Silent Mutation | TCA,TCG | S,S 66 | XP_011538682.1 | |
XM_011540381.2 | 587 | Silent Mutation | TCA,TCG | S,S 66 | XP_011538683.1 | |
XM_017016967.1 | 587 | Missense Mutation | CAG,CGG | Q,R 39 | XP_016872456.1 | |
XM_017016968.1 | 587 | Missense Mutation | CAG,CGG | Q,R 39 | XP_016872457.1 |