Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
ATGGGTTGGTGAACCAGAGGAAGGA[A/G]GTTTCTGGTCGACTGCATTGCAAAG
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 604603 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
MYOF PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
MYOF - myoferlin | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_013451.3 | 6135 | Silent Mutation | NP_038479.1 | |||
NM_133337.2 | 6135 | Silent Mutation | NP_579899.1 | |||
XM_005269693.4 | 6135 | Silent Mutation | XP_005269750.1 | |||
XM_005269694.4 | 6135 | Silent Mutation | XP_005269751.1 | |||
XM_011539632.2 | 6135 | Silent Mutation | XP_011537934.1 | |||
XM_017016068.1 | 6135 | Silent Mutation | XP_016871557.1 | |||
XM_017016069.1 | 6135 | Silent Mutation | XP_016871558.1 | |||
XM_017016070.1 | 6135 | Intron | XP_016871559.1 |