Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
GGGACAGCTCCTCCAAGCCGTCCTC[A/G]CTGTAGCCCAGTCTCTCTAGGTTGA
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 616103 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
LRIT1 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
LRIT1 - leucine rich repeat, Ig-like and transmembrane domains 1 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_015613.2 | 2018 | Silent Mutation | AGC,AGT | S,S 580 | NP_056428.1 | |
XM_011539623.2 | 2018 | Silent Mutation | AGC,AGT | S,S 531 | XP_011537925.1 | |
XM_011539626.2 | 2018 | Silent Mutation | AGC,AGT | S,S 385 | XP_011537928.1 |
LRIT2 - leucine rich repeat, Ig-like and transmembrane domains 2 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |