Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
CATTAAGGATGGCTTCACTGGCCAG[C/G]ACCTCCCTGCAGAAATAGCACAAAG
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||
Phenotype: |
MIM: 600664 MIM: 616120 MIM: 611330 | ||||||||||||||||||||
Literature Links: |
CHUK PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
CHUK - conserved helix-loop-helix ubiquitous kinase | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
CWF19L1 - CWF19-like 1, cell cycle control (S. pombe) | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001303404.1 | 1567 | Silent Mutation | GTC,GTG | V,V 453 | NP_001290333.1 | |
NM_001303405.1 | 1567 | Silent Mutation | GTC,GTG | V,V 356 | NP_001290334.1 | |
NM_001303406.1 | 1567 | Silent Mutation | GTC,GTG | V,V 356 | NP_001290335.1 | |
NM_001303407.1 | 1567 | Silent Mutation | GTC,GTG | V,V 248 | NP_001290336.1 | |
NM_018294.5 | 1567 | Silent Mutation | GTC,GTG | V,V 493 | NP_060764.3 |
SNORA12 - small nucleolar RNA, H/ACA box 12 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |