Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
AGGGGCCCGGAATCGGGAAGGACCA[A/G]TCTGCAGGTAGGAGGGCCAGCTGAA
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||
Phenotype: |
MIM: 605143 | ||||||||||||||||||||
Literature Links: |
ACTR1A PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
ACTR1A - ARP1 actin-related protein 1 homolog A, centractin alpha | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_005736.3 | 860 | Missense Mutation | ACT,ATT | T,I 251 | NP_005727.1 |
MFSD13A - major facilitator superfamily domain containing 13A | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |