Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 600466 MIM: 602141 MIM: 604592 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
ALDH3B1 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
ALDH3B1 - aldehyde dehydrogenase 3 family member B1 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
MIR4691 - microRNA 4691 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
MIR7113 - microRNA 7113 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
NDUFS8 - NADH:ubiquinone oxidoreductase core subunit S8 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_002496.3 | 267 | Missense Mutation | CAT,CGT | H,R 18 | NP_002487.1 | |
XM_005274013.1 | 267 | Missense Mutation | CAT,CGT | H,R 18 | XP_005274070.1 | |
XM_005274014.2 | 267 | Missense Mutation | CAT,CGT | H,R 18 | XP_005274071.1 | |
XM_011545053.2 | 267 | Missense Mutation | CAT,CGT | H,R 18 | XP_011543355.1 |
TCIRG1 - T-cell immune regulator 1, ATPase H+ transporting V0 subunit a3 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |