Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
GAAACCCGGCTGGAGAGGCAGCCCC[G/T]GGACTCTCCCTTCTGCGCCCCTTCC
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||
Phenotype: |
|||||||||||||||||||||
Literature Links: |
C11orf87 PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
C11orf87 - chromosome 11 open reading frame 87 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_207645.3 | 914 | Missense Mutation | CGG,CTG | R,L 136 | NP_997528.2 | |
XM_011542817.2 | 914 | Missense Mutation | CGG,CTG | R,L 283 | XP_011541119.1 | |
XM_011542818.2 | 914 | Missense Mutation | CGG,CTG | R,L 136 | XP_011541120.1 |