Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
CATTCACAACGACGAGGAGCTCTGA[G/T]TGTAATCTCTTATGATAAGAGTGTC
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||
Phenotype: |
MIM: 604102 | ||||||||||||||||||||
Literature Links: |
GRM5 PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
GRM5 - glutamate metabotropic receptor 5 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_000842.4 | 4013 | Missense Mutation | AAT,ACT | N,T 1174 | NP_000833.1 | |
NM_001143831.2 | 4013 | Missense Mutation | AAT,ACT | N,T 1206 | NP_001137303.1 | |
XM_006718828.3 | 4013 | Missense Mutation | AAT,ACT | N,T 1206 | XP_006718891.1 | |
XM_011542792.1 | 4013 | Missense Mutation | AAT,ACT | N,T 1206 | XP_011541094.1 | |
XM_017017627.1 | 4013 | Missense Mutation | AAT,ACT | N,T 1174 | XP_016873116.1 |
GRM5-AS1 - GRM5 antisense RNA 1 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |