Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
GATGTGAATTATGATCAAATCCCTT[A/G]TAATATTCACGGCATTTGTAAGGTT
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||
Phenotype: |
MIM: 605015 | ||||||||||||||||||||
Literature Links: |
ZNF214 PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
ZNF214 - zinc finger protein 214 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_013249.2 | 2142 | Silent Mutation | TAC,TAT | Y,Y 588 | NP_037381.2 | |
XM_005253128.3 | 2142 | Silent Mutation | TAC,TAT | Y,Y 588 | XP_005253185.1 | |
XM_005253129.3 | 2142 | Silent Mutation | TAC,TAT | Y,Y 574 | XP_005253186.1 | |
XM_006718308.3 | 2142 | Silent Mutation | TAC,TAT | Y,Y 588 | XP_006718371.1 | |
XM_006718309.3 | 2142 | Silent Mutation | TAC,TAT | Y,Y 574 | XP_006718372.1 | |
XM_011520355.2 | 2142 | Silent Mutation | TAC,TAT | Y,Y 599 | XP_011518657.1 | |
XM_011520357.2 | 2142 | Silent Mutation | TAC,TAT | Y,Y 574 | XP_011518659.1 | |
XM_017018269.1 | 2142 | Silent Mutation | TAC,TAT | Y,Y 574 | XP_016873758.1 |