Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
CTGCAGAAGCGGAACAGGGCCATCA[C/T]GGCCCGCAGGCAGCACCTGAAGGTA
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 607719 MIM: 191043 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
SYT8 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
SYT8 - synaptotagmin 8 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
TNNI2 - troponin I2, fast skeletal type | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001145829.1 | 57 | Missense Mutation | ACG,ATG | T,M 12 | NP_001139301.1 | |
NM_001145841.1 | 57 | Missense Mutation | ACG,ATG | T,M 12 | NP_001139313.1 | |
NM_003282.3 | 57 | Missense Mutation | ACG,ATG | T,M 12 | NP_003273.1 |