Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
GGAGCACAACAGGCCCCTCAGCCCC[A/G]ACGGGTGACACTGGGGGCATGGTCA
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||
Phenotype: |
MIM: 604985 | ||||||||||||||||||||
Literature Links: |
RBM4B PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
RBM4B - RNA binding motif protein 4B | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
SPTBN2 - spectrin beta, non-erythrocytic 2 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_006946.2 | 7909 | Silent Mutation | GTC,GTT | V,V 2361 | NP_008877.1 | |
XM_005274192.4 | 7909 | Silent Mutation | GTC,GTT | V,V 2361 | XP_005274249.1 | |
XM_005274193.3 | 7909 | Silent Mutation | GTC,GTT | V,V 2361 | XP_005274250.1 | |
XM_006718669.3 | 7909 | Silent Mutation | GTC,GTT | V,V 2368 | XP_006718732.1 | |
XM_006718671.3 | 7909 | Silent Mutation | GTC,GTT | V,V 2361 | XP_006718734.1 | |
XM_011545216.2 | 7909 | Intron | XP_011543518.1 | |||
XM_011545217.2 | 7909 | Intron | XP_011543519.1 | |||
XM_017018174.1 | 7909 | Silent Mutation | GTC,GTT | V,V 2361 | XP_016873663.1 | |
XM_017018175.1 | 7909 | Silent Mutation | GTC,GTT | V,V 2361 | XP_016873664.1 | |
XM_017018176.1 | 7909 | Silent Mutation | GTC,GTT | V,V 2361 | XP_016873665.1 | |
XM_017018177.1 | 7909 | Silent Mutation | GTC,GTT | V,V 2361 | XP_016873666.1 | |
XM_017018178.1 | 7909 | Silent Mutation | GTC,GTT | V,V 2361 | XP_016873667.1 |