Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
CTGCCAGCCAACAGCCCGGAGGCGG[C/T]GGCGGCGCAGGGCCACCCGGATGGC
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||
Phenotype: |
MIM: 604159 | ||||||||||||||||||||
Literature Links: |
SLC6A5 PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
SLC6A5 - solute carrier family 6 member 5 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001318369.1 | 332 | UTR 5 | NP_001305298.1 | |||
NM_004211.4 | 332 | Missense Mutation | GCG,GTG | A,V 20 | NP_004202.3 | |
XM_017018544.1 | 332 | Intron | XP_016874033.1 | |||
XM_017018545.1 | 332 | Intron | XP_016874034.1 |