Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
CAGACCCGATGATGGAGCGGACAAC[A/G]GACGACAGGGGTACATCCTGCAGGG
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||
Phenotype: |
MIM: 611826 | ||||||||||||||||||||
Literature Links: |
MRPL11 PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
MRPL11 - mitochondrial ribosomal protein L11 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_016050.4 | 511 | Silent Mutation | TCC,TCT | S,S 139 | NP_057134.1 | |
NM_170738.3 | 511 | Silent Mutation | TCC,TCT | S,S 113 | NP_733934.1 | |
NM_170739.3 | 511 | Silent Mutation | TCC,TCT | S,S 139 | NP_733935.1 | |
XM_017018135.1 | 511 | Silent Mutation | TCC,TCT | S,S 113 | XP_016873624.1 |