Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
AGGCCCTTCTGACCATCTCCATCCC[G/T]CCAGGCCCCAACCAGGGGCCCCACC
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 607097 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
SLC22A11 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
SLC22A11 - solute carrier family 22 member 11 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001307985.1 | 624 | Silent Mutation | CCG,CCT | P,P 75 | NP_001294914.1 | |
NM_018484.3 | 624 | Silent Mutation | CCG,CCT | P,P 75 | NP_060954.1 | |
XM_011545167.1 | 624 | UTR 5 | XP_011543469.1 |