Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
TTCCATGATACTTCTCGAAGACTGA[A/T]CTTTGAGGAAGCCAAAGAAGCCTGC
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||
Phenotype: |
|||||||||||||||||||||
Literature Links: |
C11orf88 PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
C11orf88 - chromosome 11 open reading frame 88 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
LAYN - layilin | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001258390.1 | 524 | Missense Mutation | AAC,ATC | N,I 63 | NP_001245319.1 | |
NM_001258391.1 | 524 | UTR 5 | NP_001245320.1 | |||
NM_001318799.1 | 524 | UTR 5 | NP_001305728.1 | |||
NM_178834.4 | 524 | Missense Mutation | AAC,ATC | N,I 55 | NP_849156.1 | |
XM_006718769.3 | 524 | UTR 5 | XP_006718832.1 | |||
XM_006718770.3 | 524 | UTR 5 | XP_006718833.1 |