Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
TTTGTGGTAATTGTGTCGAGGCATC[A/G]TGGGAAAAAGACTTTAGGGAGAACG
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||
Phenotype: |
MIM: 606148 MIM: 600393 | ||||||||||||||||||||
Literature Links: |
FADS1 PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
FADS1 - fatty acid desaturase 1 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_013402.4 | 1142 | Missense Mutation | ACG,ATG | T,M 447 | NP_037534.3 | |
XM_011545022.2 | 1142 | Missense Mutation | ACG,ATG | T,M 376 | XP_011543324.1 |
FEN1 - flap structure-specific endonuclease 1 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |