Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
CATGTACTTCCTCGTCACCTTTGGC[A/G]AGGGTGTGGAGCCCGCCAACCTCAA
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||
Phenotype: |
MIM: 602537 MIM: 164340 | ||||||||||||||||||||
Literature Links: |
CAPN5 PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
CAPN5 - calpain 5 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_004055.4 | 439 | Intron | NP_004046.2 | |||
XM_011545225.1 | 439 | Intron | XP_011543527.1 | |||
XM_017018223.1 | 439 | Intron | XP_016873712.1 |
OMP - olfactory marker protein | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_006189.1 | 439 | Missense Mutation | AAG,GAG | K,E 147 | NP_006180.1 |