Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
CCTCCTGGAGGCGGCCCTGGCCGCC[A/G]AGGTGAAGAAACCTGCAGCCGCAGC
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||
Phenotype: |
MIM: 600943 | ||||||||||||||||||||
Literature Links: |
SERPINH1 PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
SERPINH1 - serpin family H member 1 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001207014.1 | 169 | Intron | NP_001193943.1 | |||
NM_001235.3 | 169 | Intron | NP_001226.2 | |||
XM_011545327.1 | 169 | Missense Mutation | AAG,GAG | K,E 20 | XP_011543629.1 |