Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
CTCCAGTCATCTGTAGGCGACAAAT[A/C]GGGCAATTCCTGTGTCGATCACTCC
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 102772 MIM: 616641 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
AMPD3 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
AMPD3 - adenosine monophosphate deaminase 3 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
MIR4485 - microRNA 4485 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
MTRNR2L8 - MT-RNR2-like 8 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
RNF141 - ring finger protein 141 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_016422.3 | 720 | Silent Mutation | CCG,CCT | P,P 189 | NP_057506.2 |