Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
TTCCCACCATGCTTGCTGTGTTATG[A/G]TTGGATGCTCCAGAGATCCAGGCAA
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||
Phenotype: |
MIM: 605470 | ||||||||||||||||||||
Literature Links: |
MMP26 PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
MMP26 - matrix metallopeptidase 26 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
OR51L1 - olfactory receptor family 51 subfamily L member 1 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001004755.1 | 264 | Nonsense Mutation | TGA,TGG | *,W 88 | NP_001004755.1 |