Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
TGTGTTAGGGGAGCCAGGAAGTTAC[C/G]TGAGAAGAAACTGAGGATGGTAGTG
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||
Phenotype: |
|||||||||||||||||||||
Literature Links: |
TMEM225 PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
TMEM225 - transmembrane protein 225 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001013743.2 | 802 | Missense Mutation | CGT,GGT | R,G 110 | NP_001013765.2 | |
XM_006718832.3 | 802 | Missense Mutation | CGT,GGT | R,G 60 | XP_006718895.1 | |
XM_011542802.2 | 802 | Missense Mutation | CGA,GGA | R,G 110 | XP_011541104.1 | |
XM_017017653.1 | 802 | Missense Mutation | CTT,GTT | L,V 110 | XP_016873142.1 |