Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
ACGCGTGCCAGCTCCATGCCGGGCC[C/T]GGGAGCCGGCCAGCCACGCCATCAA
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 608965 MIM: 609849 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
CABP4 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
CABP4 - calcium binding protein 4 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001300895.1 | 175 | Intron | NP_001287824.1 | |||
NM_001300896.1 | 175 | UTR 5 | NP_001287825.1 | |||
NM_145200.3 | 175 | Intron | NP_660201.1 | |||
XM_005274114.3 | 175 | Intron | XP_005274171.2 | |||
XM_011545181.2 | 175 | Intron | XP_011543483.1 | |||
XM_011545182.2 | 175 | Intron | XP_011543484.1 | |||
XM_011545183.2 | 175 | Intron | XP_011543485.1 | |||
XM_017018025.1 | 175 | UTR 5 | XP_016873514.1 |
CORO1B - coronin 1B | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
GPR152 - G protein-coupled receptor 152 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_206997.1 | 175 | Missense Mutation | CAG,CGG | Q,R 57 | NP_996880.1 |