Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
ACCCAATGTGGCTCGCGTGGGCTCC[A/G]TGGCCATCAAGCTGTGCAATCTGCT
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 602709 MIM: 607608 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
APBB1 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
APBB1 - amyloid beta precursor protein binding family B member 1 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
SMPD1 - sphingomyelin phosphodiesterase 1 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_000543.4 | 534 | Missense Mutation | ATG,GTG | M,V 117 | NP_000534.3 | |
NM_001007593.2 | 534 | Missense Mutation | ATG,GTG | M,V 116 | NP_001007594.2 | |
NM_001318087.1 | 534 | Missense Mutation | ATG,GTG | M,V 117 | NP_001305016.1 | |
NM_001318088.1 | 534 | UTR 5 | NP_001305017.1 | |||
XM_011520303.1 | 534 | Missense Mutation | ATG,GTG | M,V 117 | XP_011518605.1 | |
XM_011520304.1 | 534 | Missense Mutation | ATG,GTG | M,V 117 | XP_011518606.1 |