Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
GCTATTGGCTCAGCTGGTTAGTGCC[A/C]TGTGCAGCAGGTACTGCACAAGCGC
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||
Phenotype: |
MIM: 609914 | ||||||||||||||||||||
Literature Links: |
AQP11 PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
AQP11 - aquaporin 11 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_173039.2 | 724 | Missense Mutation | ATG,CTG | M,L 129 | NP_766627.1 | |
XM_005273917.4 | 724 | Missense Mutation | ATG,CTG | M,L 129 | XP_005273974.1 |
LOC107987163 - uncharacterized LOC107987163 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
XM_017018644.1 | 724 | Intron | XP_016874133.1 |