Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
CTCTTGAAGGCTTTTTCCTGTGGCA[C/T]TGGCAGATGGCTGTGCTATCTTCTG
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
|||||||||||||||||||||||||||||||||||||||
Literature Links: |
C11orf65 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
C11orf65 - chromosome 11 open reading frame 65 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
KDELC2 - KDEL motif containing 2 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_153705.4 | 1527 | Silent Mutation | CAA,CAG | Q,Q 496 | NP_714916.3 | |
XM_011542621.2 | 1527 | Silent Mutation | CAA,CAG | Q,Q 482 | XP_011540923.1 | |
XM_011542623.2 | 1527 | Silent Mutation | CAA,CAG | Q,Q 358 | XP_011540925.1 |