Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
GTCCAAGGTTTTCATGACCACAGCA[C/T]AGACGCTGTCATTCACGGGAACAAA
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||
Phenotype: |
MIM: 611904 | ||||||||||||||||||||
Literature Links: |
LOC100132686 PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
LOC100132686 - uncharacterized LOC100132686 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
PLET1 - placenta expressed transcript 1 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001145024.1 | 483 | Missense Mutation | TAT,TGT | Y,C 71 | NP_001138496.1 |