Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
AGCTGGGCTCGGGGCAGGGGTCAGC[A/G]CGGGCATGGCAGGCACGGTGAATGT
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 614413 MIM: 604648 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
ACY3 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
ACY3 - aminoacylase 3 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_080658.1 | 1100 | Missense Mutation | GCG,GTG | A,V 310 | NP_542389.1 | |
XM_017018549.1 | 1100 | Missense Mutation | GCG,GTG | A,V 317 | XP_016874038.1 | |
XM_017018550.1 | 1100 | Missense Mutation | GCG,GTG | A,V 317 | XP_016874039.1 | |
XM_017018551.1 | 1100 | Missense Mutation | GCG,GTG | A,V 293 | XP_016874040.1 |
TBX10 - T-box 10 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |