Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
CTGGAACCATAGGACTTTGATTCTT[C/T]GGAGGAACTGGGACCGAGGGACCTT
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||
Phenotype: |
MIM: 601783 | ||||||||||||||||||||
Literature Links: |
MAP6 PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
MAP6 - microtubule associated protein 6 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_033063.1 | 4263 | Silent Mutation | CCA,CCG | P,P 537 | NP_149052.1 | |
NM_207577.1 | 4263 | Intron | NP_997460.1 | |||
XM_006718556.3 | 4263 | Intron | XP_006718619.1 | |||
XM_011545039.2 | 4263 | Intron | XP_011543341.1 | |||
XM_017017755.1 | 4263 | Silent Mutation | CCA,CCG | P,P 553 | XP_016873244.1 | |
XM_017017756.1 | 4263 | Silent Mutation | CCA,CCG | P,P 252 | XP_016873245.1 | |
XM_017017757.1 | 4263 | Silent Mutation | CCA,CCG | P,P 236 | XP_016873246.1 | |
XM_017017758.1 | 4263 | Silent Mutation | CCA,CCG | P,P 224 | XP_016873247.1 |