Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
GATCATGGCGGGTTTGTTGAGCTCA[G/T]TTGAATAGAATGGTACCCAAGTGTT
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||
Phenotype: |
MIM: 179616 | ||||||||||||||||||||
Literature Links: |
C11orf74 PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
C11orf74 - chromosome 11 open reading frame 74 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
RAG2 - recombination activating gene 2 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_000536.3 | 1512 | Missense Mutation | AAT,ACT | N,T 436 | NP_000527.2 | |
NM_001243785.1 | 1512 | Missense Mutation | AAT,ACT | N,T 436 | NP_001230714.1 | |
NM_001243786.1 | 1512 | Missense Mutation | AAT,ACT | N,T 436 | NP_001230715.1 |