Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
ATAGTTCTACGAAGATCAGACACTA[A/G]CTTAACAAAATTGAAATTTAAGGAA
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 603297 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
DYNC2H1 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
DYNC2H1 - dynein cytoplasmic 2 heavy chain 1 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001080463.1 | 608 | Missense Mutation | AAC,AGC | N,S 155 | NP_001073932.1 | |
NM_001377.2 | 608 | Missense Mutation | AAC,AGC | N,S 155 | NP_001368.2 | |
XM_006718903.2 | 608 | Missense Mutation | AAC,AGC | N,S 155 | XP_006718966.1 | |
XM_017018291.1 | 608 | Missense Mutation | AAC,AGC | N,S 155 | XP_016873780.1 | |
XM_017018292.1 | 608 | Intron | XP_016873781.1 | |||
XM_017018293.1 | 608 | Missense Mutation | AAC,AGC | N,S 155 | XP_016873782.1 |