Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
GTGGAGGGGGCGGCAGCAGCTGCAG[C/T]GGGCATCCACCTGCTGGGTGATGAT
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 607242 MIM: 158374 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
AP2A2 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
AP2A2 - adaptor related protein complex 2 alpha 2 subunit | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
MUC6 - mucin 6, oligomeric mucus/gel-forming | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_005961.2 | 7226 | Missense Mutation | CAC,CGC | H,R 2392 | NP_005952.2 |